Learn More
Thermo Scientific™ Owl™ Power Supply Leads, 3 ft.
Product Code. 11822684
Change view
Click to view available options
For Use With (Equipment):
Owl™ System
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
11822684 Owl™ System Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11822684 Supplier Thermo Scientific™ Supplier No. PSL5
  • ONLINE EXCLUSIVE PRICE
    41.45€
    38.70€ / pezzi 1
    Risparmia 2.75€
    6% Sconto
Spedizione stimata: 03-12-2026
Connect power supplies with Thermo Scientific™ Owl™ Power Supply Leads. These 3′ leads come with a retractable sheath end for convenience and safety.
Connect power supplies with Thermo Scientific™ Owl™ Power Supply Leads. These 3' leads come with a retractable sheath end for convenience and safety.
  • Manufactured with a strain relief cord to prevent breakage
Includes: One black and one red lead
TRUSTED_SUSTAINABILITY
Specifications
Description Power supply leads; 3 ft.
For Use With (Equipment) Owl™ System
Product Line Owl
Type Cords and Connectors
Length (English) 3 ft.