Learn More
Oxalic Acid Solution, 0.5M, Chem Lab™
Product Code. 10446932
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10446932 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10446932 Supplier Chem-Lab Supplier No. CL05.1504.1000
  • ONLINE EXCLUSIVE PRICE
    68.00€
    35.10€ / 1L
    Risparmia 32.90€
    48% Sconto
 Restricted Item
Spedizione stimata: 10-02-2027
Specifications
Density 1.02g/mL
Quantity 1 L
compliance-icons
Product Identifier
  • Oxalic acid 0.5 mol/l
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
Precautionary Statement
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
Supplemental information
  • MIXTURE LIST-Contain: Oxalic acid