Learn More
Thermo Scientific™ Oxoid™ Ampicillin/Sulbactam Antimicrobial Susceptibility discs
Codice prodotto. 11932952
Cambia vista
Click to view available options
Concentrazione:
20 μg
Dimensione della confezione:
pezzi 250
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Concentrazione unitSize
11932952 20 μg pezzi 250
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 11932952 Fornitore Thermo Scientific™ N. del fornitore CT0520B
  • ONLINE EXCLUSIVE PRICE
    59.13€
    55.05€ / pezzi 250
    Risparmia 4.08€
    6% Sconto
Visualizza tutte le promozioni
 Articolo soggetto a restrizioni
Spedizione stimata: 20-08-2026
Aggiungi al carrello
Aggiungi al carrello
Determine the susceptibility of microorganisms using manual AST methods and a full range of high-quality, easy-to-use antimicrobial susceptibility testing discs.
Manually determine the antibiotic susceptibility of microorganisms using Antimicrobial Susceptibility Testing (AST) methods in conjunction with reliable, easy-to-use Thermo Scientific™ Oxoid™ Ampicillin/Sulbactam Antimicrobial Susceptibility discs.
Ease-of-use is provided by shatterproof spring-loaded cartridges, easy-to-read labeling and end-of-cartridge indicators.
disc diffusion not only categorizes resistant, intermediate and sensitive organisms through quantitative results but also provides a visual indication of the following:
  • Inoculum level
  • Presence of contamination
  • Resistant mutants
  • Beta-lactamase activity
  • Antagonism and synergy between antibiotics
Each cartridge is individually sealed with a desiccant to maintain a moisture level of <2%, ensuring long term stability.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY
Specifica
Agenti antimicrobici Ampicillin/Sulbactam
Certificazione/Conformità Complies with CLSI, EUCAST, SFM guidelines
Concentrazione 20 μg
Descrizione Oxoid Ampicillin/Sulbactam Antimicrobial Susceptibility Discs
Codice antibiotico SAM
Product Type Antimicrobial Susceptibility Discs
Test Type Antimicrobial Susceptibility
N. per confezione 5 Cartridges/Pk.
Tipo di imballaggio 50 Discs/Cartridge
Quantità 50 Discs/Cartridge