Learn More
Thermo Scientific™ Bacillus Cereus Enterotoxin Reversed Passive Latex Agglutination Kit (BCET – RPLA)
Codice prodotto. 11498299
Cambia vista
Click to view available options
Quantità:
20 Tests/Kit
Dimensione della confezione:
20 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
11498299 20 Tests/Kit 20 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 11498299 Fornitore Thermo Scientific™ N. del fornitore TD0950A
  • ONLINE EXCLUSIVE PRICE
    565.00€
    154.47€ / 20 tests
    Risparmia 410.53€
    72% Sconto
 Articolo soggetto a restrizioni
Spedizione stimata: 08-01-2027
Rapidly detect Bacillus cereus enterotoxin (diarrhoeal type) in foods and culture filtrates with this reversed passive agglutination assay.
Rapidly detect Bacillus cereus enterotoxin (diarrhoeal type) in foods and culture filtrates with Thermo Scientific™ Oxoid™ Bacillus Cereus Enterotoxin Reversed Passive Latex Agglutination Kit. The technique of reversed passive latex agglutination (RPLA) enables soluble antigens such as bacterial toxins to be detected in an agglutination assay.
The BCET-RPLA test may be used to detect B. cereus enterotoxin in a variety of foods and to give a semi-quantitative result. The test may also be used to demonstrate enterotoxin production by isolates of B. cereus grown in culture.
Bacillus cereus is a common inhabitant of many different environments and can easily contaminate food. If contaminated foods are not cooled sufficiently after cooking and there is an extended time between preparation and consumption of foods, then surviving heat-resistant spores can germinate, enabling the organism to multiply and produce toxins. Under these circumstances, B cereus can cause food poisoning.
Use Bacillus cereus Enterotoxin RPLA test for simple and semi-quantitative detection of staphylococcal enterotoxins in food samples.
  • Simple – easy to implement
  • Visual – easy to read results
In a standard agglutination assay, the soluble antibody reacts with a particulate antigen such as bacterial cells. However, in a reversed agglutination assay the antibody, which is attached to particles, reacts with the soluble antigen. The particles (in this case, latex) do not themselves play a part in the reaction and they are therefore passive. The cross-linking of the latex particles by the specific antigen/antibody reaction results in the visible latex agglutination reaction.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY
Specifica
Content And Storage 2°C to 8°C
Descrizione BCET-RPLA Toxin Detection Kit
Tipo di imballaggio 1 Kit
Quantità 20 Tests/Kit