Learn More
p-Toluenesulfonyl Chloride, TRC
Specifica
Specifica
| Materiale o nome chimico | p-Toluenesulfonyl Chloride |
| CAS | 98-59-9 |
| Note sul grado di purezza | HPLC |
| Formula molecolare | C7H7ClO2S |
| Quantità | 250 g |
| Sinonimo | p-Toluenesulfonyl Chloride, 4-Methylbenzene-1-sulfonyl Chloride, 4-Methylbenzenesulfonyl Chloride, 4-Methylphenylsulfonyl Chloride, 4-Toluenesulfonyl Chloride, 4-Toluolsulfonyl Chloride, 4-Tosyl Chloride, NSC 175822, Toluenesulfonyl Chloride, Tosyl Chloride, p-Methylbenzenesulfonyl Chloride, p-Methylphenylsulfonyl Chloride, p-Toluenesulfochloride, p-Toluenesulfonic Acid Chloride, p-Toluenesulfonic Chloride, p-Toluenesulphonyl Chloride, p-Tosyl Chloride, TsCl, pTsCl, p-TsCl','Ts-Cl |
| InChI Formula | InChI=1S/C7H7ClO2S/c1-6-2-4-7(5-3-6)11(8,9)10/h2-5H,1H3 |
| SMILES | Cc1ccc(cc1)S(=O)(=O)Cl |
| IUPAC Name | 4-methylbenzenesulfonyl chloride |
| Molecular Weight (g/mol) | 190.65 |
| Vedi altri risultati |