Learn More
PALMD Antibody, Novus Biologicals™
Codice prodotto. 18640287 Sfoglia Tutto Bio Techne Prodotti
Cambia vista
Click to view available options
Quantità:
100 μg
50 μg
Dimensione della confezione:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
18640287 50 μg 50µL
18635138 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Codice prodotto. 18640287 Fornitore Novus Biologicals N. del fornitore NBP31259850ul
Rabbit Polyclonal Antibody
PALMD Polyclonal antibody specifically detects PALMD in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifica
Antigene PALMD
Applicazioni Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classificazione Polyclonal
Coniugato Unconjugated
Diluizione Western Blot 1:500-1:2000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50-1:200
Formulazione PBS, pH7.0
Alias gene C1orf11, palmdelphin, PALMLFLJ20271, Paralemnin-like protein
Specie ospite Rabbit
Immunogeno Produced in rabbits immunized with E. coli-derived Human PALMD fragment
Metodo di purificazione Antigen and protein A Affinity-purified
Quantità 50 μg
Status giuridico RUO
Disciplina di ricerca Apoptosis, Cancer
Primario o secondario Primary
ID gene (immissione) 54873
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Forma Purified
Isotype IgG