Learn More
PEPperCHIP™ Pan-Corona Spike Protein Microarray
Codice prodotto. 16638682
Cambia vista
Click to view available options
Quantità:
1 Slide
Dimensione della confezione:
Pezzo
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
16638682 1 Slide Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 16638682 Fornitore PEPperCHIP™ N. del fornitore SPC.052.001
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Visualizzare prodotti alternativi
Pan-Corona Spike Protein Microarray with 4,564 specific peptides of seven human coronavirus spike proteins printed in duplicate. The microarray can be used for epitope mapping, serum screening or biomarker discovery.
  • Microarray Content: Spike proteins of SARS-CoV-2, SARS-CoV, MERS-CoV, HCoV-OC43, HCoV-HKU1, HCoV-NL63 and HCoV-229E translated into overlapping peptides
  • Peptide Length/Overlap: 15 aa/ 13 aa
  • Number of Peptides: 4,564 printed in duplicate and HA and polio control peptides
  • Related Products: PEPperCHIP™ Incubation Tray 3/1, PEPperCHIP™ anti-HA control antibodies, MAPIX Analyzer
Specifica
Tipo Spike Protein Microarray
Specie Spike Proteins from SARS-CoV-2, SARS-CoV, MERS-CoV, HCoV-OC43, HCoV-HKU1, HCoV-NL63 and HCoV-229 E
Da utilizzare con (applicazione) Epitope Mapping, Serum Screening or Biomarker Discovery
Quantità 1 Slide
Research Category Infectious Disease