Learn More
Pantoprazole Sulphone N-Oxide, Mikromol™
Codice prodotto. 30052294
Cambia vista
Click to view available options
Quantità:
25 mg
100 mg
4 x 25 mg
Dimensione della confezione:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
30052294 25 mg 25mg
17933416 100 mg 100mg
30459546 4 x 25 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Codice prodotto. 30052294 Fornitore Mikromol N. del fornitore MM0099.070025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Pantoprazole Sulphone N-Oxide
TRUSTED_SUSTAINABILITY
Specifica
Materiale o nome chimico Pantoprazole Sulfone N-Oxide
CAS 953787-55-8
Nomi componenti o analita Pantoprazole Sulfone N-Oxide
Shipping Condition Room Temperature
Formula molecolare C16 H15 F2 N3 O6 S
Certificazione/Conformità ISO 17025
Sinonimo Pantoprazole Sulphone N-Oxide, 6-(Difluoromethoxy)-2-[[(3,4-dimethoxy-1-oxido-2-pyridinyl)methyl]sulfonyl]-1H-benzimidazole
InChI Formula InChI=1S/C16H15F2N3O6S/c1-25-13-5-6-21(22)12(14(13)26-2)8-28(23,24)16-19-10-4-3-9(27-15(17)18)7-11(10)20-16/h3-7,15H,8H2,1-2H3,(H,19,20)
SMILES COc1cc[n+]([O-])c(CS(=O)(=O)c2nc3cc(OC(F)F)ccc3[nH]2)c1OC
IUPAC Name 5-(difluoromethoxy)-2-[(3,4-dimethoxy-1-oxidopyridin-1-ium-2-yl)methylsulfonyl]-1H-benzimidazole
Molecular Weight (g/mol) 415.37
Quantità 25 mg
Peso formulazione 415.065 g/mol