missing translation for 'onlineSavingsMsg'
Learn More

Pantoprazole Sulphone N-Oxide, Mikromol™

Product Code. 30459546
Change view
Click to view available options
Quantità:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantità unitSize
30459546 4 x 25 mg 100mg
30052294 25 mg 25mg
17933416 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30459546 Supplier Mikromol Supplier No. MM0099.0700254AMP

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Pantoprazole Sulphone N-Oxide, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Materiale o nome chimico Pantoprazole Sulfone N-Oxide
CAS 953787-55-8
Nomi componenti o analita Pantoprazole Sulfone N-Oxide
Shipping Condition Room Temperature
Formula molecolare C16 H15 F2 N3 O6 S
Certificazione/Conformità ISO 17025
Sinonimo Pantoprazole Sulphone N-Oxide, 6-(Difluoromethoxy)-2-[[(3,4-dimethoxy-1-oxido-2-pyridinyl)methyl]sulfonyl]-1H-benzimidazole
InChI Formula InChI=1S/C16H15F2N3O6S/c1-25-13-5-6-21(22)12(14(13)26-2)8-28(23,24)16-19-10-4-3-9(27-15(17)18)7-11(10)20-16/h3-7,15H,8H2,1-2H3,(H,19,20)
Tipo di soluzione Neat
SMILES COc1cc[n+]([O-])c(CS(=O)(=O)c2nc3cc(OC(F)F)ccc3[nH]2)c1OC
IUPAC Name 5-(difluoromethoxy)-2-[(3,4-dimethoxy-1-oxidopyridin-1-ium-2-yl)methylsulfonyl]-1H-benzimidazole
Molecular Weight (g/mol) 415.37
Quantità 4 x 25 mg
Peso formulazione 415.065 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.