Learn More
Cole-Parmer™ WD-200 Series Wax Dispenser
Codice prodotto. 15804285
Cambia vista
Click to view available options
Tensione:
220/240 V
Dimensione della confezione:
Pezzo
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Tensione Dimensione della confezione
15804285 220/240 V Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 15804285 Fornitore Cole-Parmer™ N. del fornitore 8523007
  • ONLINE EXCLUSIVE PRICE
    3290.00€
    2853.09€ / pezzi 1
    Risparmia 436.91€
    13% Sconto
Spedizione stimata: 15-12-2026
Prevent wax blockage with heated tap with the WD-200 Series Wax Dispenser.
  • Accurate temperature control with digital display
  • Constructed of stainless steel with antimicrobial coating to inhibit bacteria growth
  • Enhanced safety with high temperature cut-off at 120°C
  • Fully insulated tank prevents heat loss and ensures the outer surface stays cool
This paraffin wax dispenser provides ultrafast heating for on-demand molten wax, ideal for your histology and pathology applications. The heated tap prevents blockage providing a smooth even flow of wax every time. The nondrip lever has three settings—off, on-demand pull dispense, and continuous flow. The 0.5 mm mesh filter screen prevents coarse particles from entering the tap.
Temperature can be set from ambient to 70°C using digital temperature control panel. The on/off temperature control keeps accuracy to ±1°C. The 7.5 L capacity tank can melt up to 13.2 lb (6 kg) of pelletized wax at a time. When set at 65°C, the unit will take approximately 90 minutes to fully melt 11 lb. (5 kg) of pelletized wax.
Specifica
Altezza (sistema metrico) 455 mm
Frequenza 50/60 Hz
Tensione 220/240 V
Capacità (sistema imperiale) 2 gal.
Capacità (sistema metrico) 7.5 L
Descrizione Paraffin Wax Dispenser
Modello MH8524