missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PARN Polyclonal specifically detects PARN in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigene | PARN |
| Applicazioni | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classificazione | Polyclonal |
| Coniugato | Unconjugated |
| Diluizione | Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Formulazione | PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide |
| Alias gene | DANEC 3.1.13.4, Deadenylating nuclease, Deadenylation nuclease, poly(A)-specific ribonuclease (deadenylation nuclease), poly(A)-specific ribonuclease PARN, Polyadenylate-specific ribonuclease |
| Simboli geni | PARN |
| Specie ospite | Rabbit |
| Immunogeno | This antibody was developed against Recombinant Protein corresponding to amino acids:DGEFSGISDGPSVSALTNGFDTPEERYQKLKKHSMDFLLFQFGLCTFKYDYTDSKYITKSFNFYVFPKPFNRSSPDVKFVCQSSSIDFLASQGFDFNKVFRNGIPYLNQEEERQLREQYDEKRSQANGAGA |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?