missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX PathHunter™ CCR6 Activated GPCR Internalization U2OS Cell Line
Descrizione
The PathHunter™ CCR6 Activated GPCR internalization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) affecting GPCR activation and internalization. This cell line enables assessment of ligand (e.g. CCL20) based internalization of CCR6 via detection of localization to the early endosome. This product uses EFC technology. The included cell line overexpresses EA-tagged β-arrestin 2 as well as a PK-tagged early endosome protein and untagged CCR6. Upon ligand-induced receptor activation, β-arrestin 2 is recruited to the activated GPCR and the resulting CCR6-arrestin complex is internalized to the early endosome, leading to production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells.
Specifica
Specifica
| Assay Entry | Receptor Internalization |
| Linea cellulare | U2OS |
| Da utilizzare con (applicazione) | Cell Lines |
| Primary Agonist | Recombinant Human MIP-3α (CCL20) |
| Quantità | 2 Vials |
| Target Entry | CCR6 |
| Numero di accesso | AY242126 |
Titolo del prodotto
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Individuate un'opportunità di miglioramento?