missing translation for 'onlineSavingsMsg'
Learn More

DiscoverX PathHunter™ CHO-K1 mCRHR1 β-Arrestin Cell Line

Codice prodotto. 17383239
Encompass
Cambia vista
Click to view available options
Quantità:
2 Vials
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità
17383239 2 Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 17383239 Fornitore DiscoverX N. del fornitore 930473C2

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

The PathHunter™ CHO-K1 mCRHR1 β-Arrestin Cell Line measures mCRHR1 (GPCR) activity via recruitment of β-Arrestin2. This cell line ships as two vials of cryopreserved cells. Mouse protein.

The PathHunter™ mCRHR1 β-Arrestin cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. CRH, sauvagine) based activation of mCRHR1 GPCR activity via detection of β-arrestin recruitment. This product uses EFC technology (www.discoverx.com/technologies-platforms/enzyme-fragment-complementation-technology). The included cell line overexpresses PK-tagged mCRHR1 and EA-tagged β-Arrestin 2. Activation of mCRHR1 stimulates the recruitment of β-Arrestin 2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. Please note that this product expresses the Mouse version of this protein.

Specifica

Assay Entry β-Arrestin recruitment
Linea cellulare CHO-K1
Da utilizzare con (applicazione) Cell Lines
Primary Agonist Sauvagine
Quantità 2 Vials
Target Entry mCRHR1
Numero di accesso NM_007762
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.