missing translation for 'onlineSavingsMsg'
Learn More

DiscoverX PathHunter™ eXpress mGIPR U2OS β-Arrestin GPCR Assay

Codice prodotto. 17363889
Encompass
Cambia vista
Click to view available options
Quantità:
1,000 dp (10 x 96-well)
Dimensione della confezione:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
17363889 1,000 dp (10 x 96-well) 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 17363889 Fornitore DiscoverX N. del fornitore 930846E3MCP5L

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

The PathHunter™ eXpress mGIPR U2OS β-Arrestin GPCR Assay measures mGIPR (GPCR) activity via recruitment of β-Arrestin2. eXpress kits contain all assay materials: cells, reagents, and plates. Mouse version.

The PathHunter™ mGIPR β-Arrestin eXpress Kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. GIP) based activation of mGIPR GPCR activity via detection of β-arrestin recruitment. This product uses EFC technology (www.discoverx.com/technologies-platforms/enzyme-fragment-complementation-technology). The included cells overexpress PK-tagged mGIPR and EA-tagged β-Arrestin 2. Activation of mGIPR stimulates the recruitment of β-Arrestin 2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. Please note that this product expresses the Mouse version of this protein.

Specifica

Assay Entry GPCR
Da utilizzare con (applicazione) Assay-Ready eXpress Kits
Quantità 1,000 dp (10 x 96-well)
Target Entry mGIPR
Linea cellulare U2OS
Bersaglio mGIPR
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.