missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX PathHunter™ U2OS EphA4 Functional Assay
Descrizione
The PathHunter™ U2OS EphA4 Functional Assay provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting RTKs. This kit assesses RTK activation via detection of activated EphA4 dimerization-induced SH2 domain protein recruitment. This product uses EFC technology. The included cell line overexpresses PK-tagged EphA4 and an EA-tagged SH2 domain protein. Upon ligand-induced receptor activation, SH2-EA recruitment leads to production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells.
Specifica
Specifica
| Assay Entry | U2OS |
| Linea cellulare | U2OS |
| Da utilizzare con (applicazione) | Cell Lines |
| Primary Agonist | EphrinA3 |
| Quantità | 2 Vials |
| Target Entry | EphA4 |
| Numero di accesso | NM_004438.3 |
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?