missing translation for 'onlineSavingsMsg'
Learn More

DiscoverX PathHunter™ U2OS ErbB2-ErbB3 Functional Assay

Codice prodotto. 17303359
Encompass
Cambia vista
Click to view available options
Quantità:
2 Vials
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità
17303359 2 Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 17303359 Fornitore DiscoverX N. del fornitore 930535C3

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

The PathHunter™ U2OS ErbB2-ErbB3 Functional Assay measures ErbB2-ErbB3 receptor tyrosine kinase activity via ligand-induced SH2 domain protein recruitment. This cell line ships as two vials of cryopreserved cells.

The PathHunter™ U2OS ErbB2-ErbB3 Functional Assay provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting RTKs. This kit assesses RTK activation via detection of activated ErbB2 and ErbB3 dimerization-induced SH2 domain protein recruitment. This product uses EFC technology. The included cell line overexpresses PK-tagged ErbB2 and an EA-tagged SH2 domain protein. It also overexpresses an untagged coreceptor, ErbB3 Upon ligand-induced receptor activation, SH2-EA recruitment leads to production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells.

Specifica

Assay Entry Receptor Activation
Linea cellulare U2OS
Da utilizzare con (applicazione) Cell Lines
Primary Agonist Recombinant Human Heregulinβ-1
Quantità 2 Vials
Target Entry ErbB2-ErbB3
Numero di accesso NM_004448.2
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.