missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX PathHunter™ U2OS mRHO(P23H) MEM-EA Pharmacotrafficking Assay
Descrizione
The PathHunter™ U2OS mRHO(P23H) MEM-EA Pharmacotrafficking Assay is a stable, engineered cell line for use in studying pharmacochaperones targeting mRHO. This cell line enables assessment of chaperone-based stabilization of mRHO(P23H) via detection of rescued membrane trafficking. This product uses EFC technology. The included cell line overexpresses PK-tagged mutant mRHO(P23H), which is misfolded and retained in the ER. The cell line also expresses membrane-localized EA. When a pharmacochaperone stabilizes the mutant mRHO, it is trafficked to the membrane, producing EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells.
Specifica
Specifica
| Assay Entry | Pharmacotrafficking |
| Linea cellulare | U2OS |
| Da utilizzare con (applicazione) | Cell Lines |
| Primary Agonist | 9-cis-Retinal |
| Quantità | 2 Vials |
| Target Entry | mRHO(P23H) |
| Numero di accesso | NM_145383.1 |
Titolo del prodotto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?