Learn More
DiscoverX PathHunter™ U2OS OPRK1 β-Arrestin Cell Line
Descrizione
The PathHunter™ OPRK1 β-Arrestin cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. dynorphin A) based activation of OPRK1 GPCR activity via detection of β-arrestin recruitment. This product uses EFC technology (www.discoverx.com/technologies-platforms/enzyme-fragment-complementation-technology). The included cell line overexpresses PK-tagged OPRK1 and EA-tagged β-Arrestin 2. Activation of OPRK1 stimulates the recruitment of β-Arrestin 2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells.
Specifica
Specifica
| Linea cellulare | U2OS |
| Da utilizzare con (applicazione) | Cell Lines |
| Primary Agonist | Dynorphin A |
| Quantità | 2 Vials |
| Target Entry | OPRK1 |
| Numero di accesso | AF498922 |