missing translation for 'onlineSavingsMsg'
Learn More

DiscoverX PathHunter™ U2OS PRβ Protein Interaction Cell Line

Codice prodotto. 17323899
Encompass
Cambia vista
Click to view available options
Quantità:
2 Vials
Dimensione della confezione:
2 fiale
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
17323899 2 Vials 2 fiale
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 17323899 Fornitore DiscoverX N. del fornitore 930851C3

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

The PathHunter™ U2OS PRβ Protein Interaction Cell Line measures activated PRβ nuclear hormone receptor interaction with SRCPs. This cell line ships as two vials of cryopreserved cells.

The PathHunter™ PRβ Protein Interaction cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) affecting NHR activation and recruitment of cofactors. This cell line enables assessment of ligand (e.g. testosterone, norgestrel) based NHR activation via detection of SRCP (steroid receptor co-activator peptide) recruitment. This product uses EFC technology. The included cell line overexpresses PK-tagged PRβ NHR and EA-tagged steroid receptor co-activator peptide (SRCP). Upon activation of PRβ, induced interaction with SRCP leads to production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells.

Specifica

Assay Entry Protein Interaction
Linea cellulare U2OS
Da utilizzare con (applicazione) Cell Lines
Primary Agonist D(-)-Norgestrel
Quantità 2 Vials
Target Entry PRβ
Numero di accesso NM_000926
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.