Learn More
Gilac™ Patient Drinking Cup, Green
Product Code. 17928661
Change view
Click to view available options
Capacity (Metric):
0.25 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
17928661 0.25 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17928661 Supplier Gilac™ Supplier No. G128408
  • ONLINE EXCLUSIVE PRICE
    3.08€
    1.99€ / pezzi 1
    Risparmia 1.09€
    35% Sconto
Spedizione stimata: 09-12-2026
Patient Drinking Cup has an integrated lid and drinking straw.
  • Patient drinking cup with graduations (in mL).
  • Integrated lid and drinking straw.
  • Allows drinking without spilling content.
Specifications
Material Polypropylene
Capacity (Metric) 0.25 L
Capacity (English) 8.5 oz.
Product Type Tumbler Cup