missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PCDHGC3 Monoclonal Antibody (926531)
GREENER_CHOICE

Codice prodotto. 30534810
Cambia vista
Click to view available options
Quantità:
100 μg
Dimensione della confezione:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
30534810 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 30534810 Fornitore Invitrogen™ N. del fornitore MA561941
 Articolo soggetto a restrizioni
Spedizione stimata: 12-08-2026
Aggiungi al carrello
Aggiungi al carrello
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Mouse Monoclonal Antibody

Reconstitute at 0.5 mg/mL in sterile PBS.

This gene is a member of the protocadherin gamma gene cluster, one of three related clusters tandemly linked on chromosome five. These gene clusters have an immunoglobulin-like organization, suggesting that a novel mechanism may be involved in their regulation and expression. The gamma gene cluster includes 22 genes divided into 3 subfamilies. Subfamily A contains 12 genes, subfamily B contains 7 genes and 2 pseudogenes, and the more distantly related subfamily C contains 3 genes. The tandem array of 22 large, variable region exons are followed by a constant region, containing 3 exons shared by all genes in the cluster. Each variable region exon encodes the extracellular region, which includes 6 cadherin ectodomains and a transmembrane region. The constant region exons encode the common cytoplasmic region. These neural cadherin-like cell adhesion proteins most likely play a critical role in the establishment and function of specific cell-cell connections in the brain. Alternative splicing has been described for the gamma cluster genes.
TRUSTED_SUSTAINABILITY

Specifica

Antigene PCDHGC3
Applicazioni Immunohistochemistry (Paraffin)
Classificazione Monoclonal
Clone 926531
Concentrazione 0.5 mg/mL
Coniugato Unconjugated
Formulazione PBS with trehalose and no preservative
Gene PCDHGC3
N. accesso geni Q9UN70
Alias gene Cadherin like 2; cadherin-like 2; PC 43; PC43; PC-43; PCDH 2; PCDH gamma C3; PCDH2; PCDH-GAMMA-C3; Pcdhgb6; PCDHGC 3; PCDHGC3; protocadherin 2; protocadherin 43; protocadherin gamma subfamily C, 3; protocadherin gamma-C3; Protocadherin2; Protocadherin-2; Protocadherin43; protocadherin-43
Simboli geni PCDHGC3
Specie ospite Mouse
Immunogeno Chinese Hamster Ovary cell line CHO-derived human Protocadherin gamma C3 protein, Ser30-Tyr693.
Metodo di purificazione Protein A/G
Quantità 100 μg
Status giuridico RUO
Primario o secondario Primary
ID gene (immissione) 5098
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Forma Lyophilized
Isotype IgG1
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.