Learn More
PCPTP1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA567143
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
Immunogen sequence: ERFQLSLRQDK EKNQEIHLSP ITLQPALSEA KTVHSMVQPE QAPKVLNVVV DPQGRGAPEI KATTATSVCP SPFK Highest antigen sequence identity to the following orthologs - mouse 80%, rat 80%.
PCPTP1 Polyclonal antibody specifically detects PCPTP1 in Human samples. It is validated for Immunocytochemistry.Specifica
PCPTP1 | |
Polyclonal | |
Unconjugated | |
PTPRR | |
Ch-1 PTPase; ch-1PTPase; ECPPT; ECPTP; EC-PTP; Gmcp1; mPTP213; NC-PTPCOM1; PC12-PTP1; PCPTP1; Phosphotyrosine phosphatase 13; protein tyrosine phosphatase Cr1PTPase; protein tyrosine phosphatase PCPTP1; protein tyrosine phosphatase, receptor type R; protein tyrosine phosphatase, receptor type, R; protein-tyrosine phosphatase NC-PTPCOM1; protein-tyrosine phosphatase PCPTP1; Protein-tyrosine-phosphatase SL; protein-tyrosine-phosphatase-SL; Ptp; Ptp13; PTPBR7; PTPRQ; PTPRR; PTP-SL; receptor-type tyrosine-protein phosphatase R; R-PTP-R; RPTPRR; STEP-like PTPase; tyrosine phosphatase CBPTP | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5801 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q15256 | |
PTPRR | |
Recombinant Human PCPTP1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only