missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ PDPK1 (Human) Recombinant Protein

Human PDPK1 full-length ORF ( AAH33494.1, 1 a.a. - 429 a.a.) recombinant protein with GST-tag at N-terminal.

Marca:  Abnova™ H00005170-P01.10ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16158891

  • 302.00€ / 10µg


Spedizione stimata
26-11-2020
Aggiungi al carrello

Descrizione

Descrizione

  • Sequence: MARTTSQLYDAVPIQSSVVLCSCPSPSMVRTQTESSTPPGIPGGSRQGPAMDGTAAEPRPGAGSLQHAQPPPQPRKKRPEDFKFGKILGEGSFSTVVLARELATSREYATRANSFVGTAQYVSPELLTEKSACKSSDLWALGCIIYQLVAGLPPFRAGNEYLIFQKIIKLEYDFPEKFFPKARDLVEKLLVLDATKRLGCEEMEGYGPLKAHPFFESVTWENLHQQTPPKLTAYLPAMSEDDEDCYGNYDNLLSQFGCMQVSSSSSSHSLSASDTGLPQRSGSNIEQYIHDLDSNSFELDLQFSEDEKRLLLEKQAGGNPWHQFVENNLILKMGPVDKRKGLFARRRQLLLTEGPHLYYVDPVNKVLKGEIPWSQELRPEAKNFKTFFVHTPNRTYYLMDPSGNAHKWCRKIQEVWRQRYQSHPDAAVQ
Specifica

Specifica

AAH33494.1
3-phosphoinositide dependent protein kinase-1
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
PDPK1
GST
5170
Wheat germ expression system
10 μg
MGC20087, MGC35290, PDK1, PRO0461
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Videos
SDS
Documenti