Learn More
PDSS2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA556231
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Immunogen sequence: GDFLLANACN GLALLQNTKV VELLASALMD LVQGVYHENS TSKESYITDD IGISTWKEQT FLSHGALLAK SCQAAMELAK HDAEVQNMAF QYGKH Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 85%.
PDSS2 Polyclonal antibody specifically detects PDSS2 in Human, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifica
PDSS2 | |
Polyclonal | |
Unconjugated | |
PDSS2 | |
5430420P03Rik; All-trans-decaprenyl-diphosphate synthase subunit 2; bA59I9.3; C6orf210; Candidate tumor suppressor protein; COQ10D3; decaprenyl diphosphate synthase subunit 2; decaprenyl pyrophosphate synthase subunit 2; decaprenyl pyrophosphate synthetase subunit 2; decaprenyl-diphosphate synthase subunit 2; DLP1; EGK_15212; Gm60; hDLP1; hypothetical protein LOC436624; kd; mDLP1; Pdss2; Plmp; prenyl (decaprenyl) diphosphate synthase, subunit 2; prenyl (solanesyl) diphosphate synthase, subunit 2; prenyl diphosphate synthase, subunit 2; prenyltransferase-like mitochondrial protein; RP11-59I9.3; solanesyl-diphosphate synthase subunit 2; subunit 2 of decaprenyl diphosphate synthase; zgc:92156 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
365592, 57107 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5U2R1, Q86YH6 | |
PDSS2 | |
Recombinant protein corresponding to Human PDSS2 | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Research Use Only