Learn More
- Home
- Products
- Chromatography Supplies
- Chromatography Tubing
- Thermo Scientific™ PEEK Capillary Tubing
Thermo Scientific™ PEEK Capillary Tubing
Product Code. 10766524 Shop All Thermo Scientific Products| Product Code. | Overall Length (Metric) | Diameter (English) Inner | Color | Unit Size |
|---|---|---|---|---|
| 10766524 | 50 cm | 0.005 in. | Red | Each |
| 10259914 | 10 cm | 0.003 in. | Natural | Each |
| 10280195 | 100 cm | 0.003 in. | Natural | Each |
| 10302185 | 20 cm | 0.003 in. | Natural | Each |
| 10794267 | 30 cm | 0.003 in. | Natural | Each |
| 10006604 | 5 cm | 0.003 in. | Natural | Each |
| 10678515 | 50 cm | 0.003 in. | Natural | Each |
| 10552265 | 10 cm | 0.005 in. | Red | Each |
| 10609993 | 20 cm | 0.005 in. | Red | Each |
| 10069382 | 30 cm | 0.005 in. | Red | Each |
| 10573225 | 5 cm | 0.005 in. | Red | Each |
| 10498084 | 100 cm | 0.007 in. | Yellow | Each |
| 10594665 | 30 cm | 0.007 in. | Yellow | Each |
| 10439244 | 50 cm | 0.007 in. | Yellow | Each |
| 10504255 | 100 cm | 0.010 in. | Blue | Each |
| 10574855 | 30 cm | 0.010 in. | Blue | Each |
| 10752917 | 5 cm | 0.010 in. | Blue | Each |
| 10076304 | 50 cm | 0.010 in. | Blue | Each |
| 10581305 | 100 cm | 0.020 in. | Orange | Each |
| 10161033 | 20 cm | 0.020 in. | Orange | Each |
| 10464793 | 30 cm | 0.020 in. | Orange | Each |
| 10180553 | 50 cm | 0.020 in. | Orange | Each |
Eseguire il login per visualizzare lo stock disponibile
Descrizione
Use Thermo Scientific™ Rheodyne™ Sample Loops for your Rheodyne HPLC sample injectors. They are available in stainless steel or biocompatible PEEK.
- Broad chemical compatibility
- Biocompatible
- Easily cut to desired length
- Appropriate for many HPLC applications
- Resistant to most organic solvents however nitric acid, sulfuric acid, dichloromethane, THF and DMSO are not recommended
Specifica
Specifica
| For Use With | Rheodyne HPLC Sample Injectors |
| Type | Capillary Tubing |
| Color | Red |
| Diameter (English) Inner | 0.005 in. |
| Diameter (Metric) Inner | 0.127 mm |
| Overall Length (English) | 1 ft. 8 in. |
| Overall Length (Metric) | 50 cm |
| Material | PEEK |