Learn More
Idex Flangeless Male Nut, Standard Knurl, Natural PEEK, 1/8 in. OD Tubing, 1/4-28 Flat-Bottom
Product Code. 11795908
Change view
Click to view available options
Packaging Quantity:
10 Pk.
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Packaging Quantity Unit Size
11795908 10 Pk. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11795908 Supplier Idex Supplier No. 0200766
  • ONLINE EXCLUSIVE PRICE
    83.70€
    51.13€ / pezzi 10
    Risparmia 32.57€
    38% Sconto
Spedizione stimata: 18-12-2026
Eliminate the need to flange tubing with this Standard Knurl Flangeless Male Nut.
  • Removable and reusable—saving you money.
  • Just slip the nut and ferrule over the tubing and finger tighten the assembly into your receiving port.
  • Unlike the time required to flange tubing, it only takes a few seconds to change the components.
  • Select from various ferrule materials to suit your application.
Specifications
Type Nut
Color Natural
Packaging Quantity 10 Pk.
Thread Size For 1/8 in. 1/4 to 28 Thread Size
Material PEEK