Learn More
PEG3 Antibody, Novus Biologicals™
Codice prodotto. 18181918 Sfoglia Tutto Bio Techne Prodotti
Cambia vista
Click to view available options
Quantità:
0.1 mL
Dimensione della confezione:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
18181918 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 18181918 Fornitore Novus Biologicals N. del fornitore NBP247624
Rabbit Polyclonal Antibody
PEG3 Polyclonal specifically detects PEG3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifica
Antigene PEG3
Applicazioni Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence
Classificazione Polyclonal
Coniugato Unconjugated
Diluizione Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50
Formulazione PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
N. accesso geni Q9GZU2
Alias gene DKFZp781A095, KIAA0287PW1, paternally expressed 3, paternally expressed gene 3, Zinc finger and SCAN domain-containing protein 24, ZNF904, ZSCAN24Kruppel-type zinc finger protein
Simboli geni PEG3
Specie ospite Rabbit
Immunogeno This antibody was developed against Recombinant Protein corresponding to amino acids:SSNVREYQKARAKKKYIEHRSNETSVIHSLPFGEQTFRPRGMLYECQECGECFAHSSDLTEHQKIHDREKPSGSRNYEWSVIRSLAPTDPQTSYAQEQYAKEQARNKCKDFRQFFATSEDLNTNQKIYDQ
Metodo di purificazione Affinity Purified
Quantità 0.1 mL
Status giuridico RUO
Primario o secondario Primary
ID gene (immissione) 5178
Test di specificità Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
For Research Use Only