Learn More
3M™ PELTOR™ Alert FM-Radio Headset
Codice prodotto. 15739997
Cambia vista
Click to view available options
Product Type:
Fortified Safety Helmet, Radio Headset
Radio Headset
Modello:
Fortified Safety Helmet
Headband
Peso (sistema metrico):
400 g
430 g
Dimensione della confezione:
Pezzo
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Product Type Modello Peso (sistema metrico) unitSize
15739997 Fortified Safety Helmet, Radio Headset Fortified Safety Helmet 430 g Pezzo
15779987 Radio Headset Headband 400 g Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Codice prodotto. 15739997 Fornitore 3M™ N. del fornitore 7000108538
  • ONLINE EXCLUSIVE PRICE
    327.00€
    167.98€ / pezzi 1
    Risparmia 159.02€
    48% Sconto
Spedizione stimata: 09-09-2026
Aggiungi al carrello
Aggiungi al carrello
3M™ PELTOR™ Alert FM-Radio Headset with built in FM radio and a level dependent function to improve safety and increase motivation for wearers in noisy conditions.
  • Protect your hearing and listen to the radio - at the same time
  • Excellent FM-radio reception
  • Level dependent function lets the user hear surrounding noise and converse with colleagues
  • 3.5 mm stereo audio input plug to connect to external devices
  • Operating time approx. 70 hours
  • Flex system means you only need replace the cable if you update or change your radio manufacturer, not the expensive part, the headset
TRUSTED_SUSTAINABILITY
Specifica
Product Type Fortified Safety Helmet, Radio Headset
Modello Fortified Safety Helmet
Livello di rumorosità 30 dB
Peso (sistema metrico) 430 g
Certificazione/Conformità CE Approved