Description
- Provides reliable hearing protection in environments with moderate noise levels.
- Colour-coded yellow for easy identification within the PELTOR™ X Series.
- Electrically insulated twin headband design reduces heat build-up and ensures comfort.
- Headband features durable stainless-steel wire coated in plastic for strength and comfort.
- Flat, wide headband with soft plastic coating delivers stable, balanced pressure on the head.
- Dielectric construction with no exposed metal offers protection in electrically sensitive environments.
- Smooth-sliding adjustment points for easy fitting and repositioning.
- Wide, soft foam cushions enhance user comfort during extended wear.
- Slim cup design improves compatibility with other PPE, including hard hats and safety glasses.
- Helmet-mounted versions available as dielectric (P5) or with metal arms (P3) depending on electrical insulation requirements.
- Replaceable cushions and foam inserts (Hygiene Kits) help extend product life and maintain hygiene.
Spécification
Spécification
| Modello | X2P3E |
| Noise Reduction Rating (NRR) | 31 dB |
| Corded | No |
| Product Type | Helmet-mounted Ear Muff |
| Monouso | Reusable |
| Metal Detectable | Yes |
| Headband Color | Orange |
| Dielectric | No |
| Stile | Over-the-head |
| Foldable | Yes |
Nom du produit
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
Vous avez repéré une opportunité d'amélioration ?