Learn More
3M™ PELTOR™ Replacement Suspensions G2C
Product Code. 15770018 Shop All 3M Products
Change view
Click to view available options
Color: Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15770018 Color: Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15770018 Supplier 3M™ Supplier No. 7100002034
  • ONLINE EXCLUSIVE PRICE
    232.00€
    182.24€ / pezzi 20
    Risparmia 49.76€
    21% Sconto
Spedizione stimata: 05-02-2027
3M™ PELTOR™ Replacement Suspensions G2C is a replacement pinlock suspension with plastic sweatband for all 3M™ Hard Hats.
  • Replacement pinlock suspension cradle with plastic sweatband
  • Pinlock adjustment system providing simple and secure size adjustment for head sizes from 54 to 62 cm
  • Snaps easily into place, to provide all the comfort and easy adjustability of the original system
  • 3M™ provides reliable head and face protection products that meet regulated performance and safety requirements
  • Suitable for use with all 3M™ Hard Hats
TRUSTED_SUSTAINABILITY
Specifications
Material Plastic
Size 54 to 62 cm