Learn More
Thermo Scientific Chemicals Peptone, from meat (pork gelatine)
Sfoglia Tutto Thermo Scientific Chemicals ProdottiDescrizione
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
● Peptone, from meat, is a growth-promoting nutrient used in bacteria and fungi cultures
Applications
● It is commonly used in animal cell culture.
● Peptone is used in mass scale organism cultivation, and fermentation processes
● Peptone, from meat, is used as a supplement to in vitro culture media to help to increase cell number, specific monoclonal antibody production, and maintain long-term cultures
● This compound has been used as a component in Ax-2 axenic medium to induce reproduction of Dictyostelium discoideum
Specifica
Specifica
| Materiale o nome chimico | Peptone |
| Quantità | 250 g |
| Informazioni di solubilità | Solubility aq. soln.: soluble. |
RUO – Research Use Only
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.