Learn More
Perfluorooctanoic acid 13C4 (1,2,3,4-13C4) 50 μg/mL in Methanol:Water, Dr. Ehrenstorfer
Codice prodotto. 30327286
Cambia vista
Click to view available options
Quantità:
1 mL
Dimensione della confezione:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
30327286 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 30327286 Fornitore Dr. Ehrenstorfer N. del fornitore DREA15987154MW50
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifica
Materiale o nome chimico Perfluorooctanoic Acid 13C4 (1,2,3,4-13C4)
CAS 960315-48-4
Solvente o matrice Methanol/Water
Formula molecolare 13C4 C4 H F15 O2
Quantità 1 mL
Sinonimo 13C4-PFOA, Perfluorooctanoic acid 13C4 (1,2,3,4-13C4), Octanoic-1,2,3,4-13C4 acid, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluoro- (ACI), 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-Pentadecafluorooctanoic-1,2,3,4-13C4 acid (ACI), DTXSID 90894099, Perfluoro-n-[1,2,3,4-13C4]-octanoic acid
InChI Formula InChI=1S/C8HF15O2/c9-2(10,1(24)25)3(11,12)4(13,14)5(15,16)6(17,18)7(19,20)8(21,22)23/h(H,24,25)/i1+1,2+1,3+1,4+1
SMILES O[13C](=O)[13C](F)(F)[13C](F)(F)[13C](F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
IUPAC Name 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluoro(1,2,3,4-^{13}C_{4})octanoic acid
Molecular Weight (g/mol) 418.039
Peso formulazione 417.987 g/mol