missing translation for 'onlineSavingsMsg'
Learn More

ELAREM™ Perform-FD PLUS I GMP Grade

Codice prodotto. 30048946
Cambia vista
Click to view available options
Quantità:
50 mL
100 mL
500 mL
Dimensione della confezione:
100mL
500mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
30048946 50 mL 50mL
30048947 100 mL 100mL
30048945 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 30048946 Fornitore ELAREM™ N. del fornitore PE30621

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

GMP-grade, xeno-free cell culture supplement for safe and efficient cell expansion. Usable for a variety of cell types including MSCs, iPSCs, NK cells, dendritic cells and more. No anticoagulant added or required.

ELAREM™ Perform-FD PLUS GMP Grade is a fibrinogen-depleted and anticoagulant-free Human Platelet Lysate of EU origin. As a xeno-free and easy-to-use cell culture supplement. ELAREM™ Perform-FD PLUS supports the efficient and safe in vitro expansion of various primary cells and cell lines. ELAREM™ Perform-FD PLUS GMP Grade is suited to cell culture applications that are subject to regulatory requirements bu do not require viral inactivation steps.

  • Due to a unique fibrinogen-depletion technique, ELAREM™ Perform-FD PLUS is a clot-free and anticoagulant-free Human Platelet Lysate. As a result, the product neither requires the addition of anticoagulants in cell culture nor during manufacturing. That makes ELAREM™ Perform-FD PLUS a ready-to-use Human Platelet Lysate that is widely applicable, since the absence of anticoagulants offers even more convenient cell growth conditions.
  • ELAREM™ Perform-FD PLUS is manufactured under an ISO 9001:2015 certified Quality Management System. Each batch is produced from large pools of platelet units. That ensures batch-to-batch consistency and enables reproducible conditions. ELAREM™ Perform-FD PLUS is manufactured from platelet units obtained from healthy blood donors at licensed blood centres in the EU.

Specifications

Forma Liquid
Product Type Cell Culture Supplement
Sterilità Sterile-filtered
Sterilization Method Sterile-filtered
Quantità 50 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.