Learn More
Brady™ Permanent Paint
Product Code. 11791356 Shop All Brady Products
Change view
Click to view available options
Color:
Black
Blue
Gray
Green
Orange
Red
White
Yellow
Unit Size:
1 pack
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
11791356 Blue 1 pack
11721366 Black 1 pack
11701366 Green 1 pack
11731366 Gray 1 pack
11711366 Orange 1 pack
11761366 Red 1 pack
11751366 White 1 pack
11741366 Yellow 1 pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 11791356 Supplier Brady™ Supplier No. 225005
  • ONLINE EXCLUSIVE PRICE
    370.00€
    333.76€ / 1 confezioni
    Risparmia 36.24€
    9% Sconto
 Restricted Item
Spedizione stimata: 14-12-2026
Permanent Paint is ideal for floor marking.
  • Floor marking paint.
Specifications
Quantity 6 Pk.
Alternate Part Number EASYLINE PAINT BLUE (PAC of 6)
Product Type Paint
For Use With (Application) Floor marking
Color Blue