Learn More
DiversifiedBiotech PetriStickers™
Codice prodotto. 30282323
Cambia vista
Click to view available options
Include:
100 Square Grid
12 Sector Pie
16 Sector Pie
32 Square Grid
50 Square Grid
6 Sector Pie
70 Square Grid
8 Sector Pie
Dimensione della confezione:
pezzi 36
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Include Dimensione della confezione
30282323 50 Square Grid pezzi 36
30282301 32 Square Grid pezzi 36
30282330 70 Square Grid pezzi 36
30282337 100 Square Grid pezzi 36
30282281 6 Sector Pie pezzi 36
30282289 8 Sector Pie pezzi 36
30282343 12 Sector Pie pezzi 36
30282342 16 Sector Pie pezzi 36
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Codice prodotto. 30282323 Fornitore DiversifiedBiotech N. del fornitore PSTK1000
  • ONLINE EXCLUSIVE PRICE
    29.45€
    27.86€ / pezzi 36
    Risparmia 1.59€
    5% Sconto
Spedizione stimata: 04-11-2026
Aggiungi al carrello
Edge
Aggiungi al carrello
Allow for petri dishes to be labeled quickly, neatly and reliably
  • PetriStickers™ allow for petri dishes to be labeled quickly, neatly and reliably
  • Made with water-resistant ink, substrate and adhesives and will remain firmly attached at temperatures from 42° to 4°C
  • Simply remove the label's protective backing and apply it to the back of a petridish
  • After labeling, the dish can be held in a comfortable position for streaking or colony transfer, stored in an incubator or refrigerator, and photographed
Specifica
Tipo Petri Dish Label
Include 50 Square Grid
Diametro (sistema metrico) 7.62 cm