missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Petrol, puriss., boiling point 100° to 140°C, Honeywell
Click to view available options
Quantità:
1 L
2.5 L
25 L
Descrizione
- We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Specifica
Materiale o nome chimico | Petrol |
Densità | 0.720 to 0.745g/mL at 20°C |
Punti di ebollizione | 100°C to 140°C |
Punto d'infiammabilità | <10°C (<50°F) |
Quantità | 1 L |
Numero UN | 3295 |
Grado | puriss. |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'