Learn More
PF 04457845, Tocris Bioscience™
Potent and selective irreversible FAAH inhibitor
Marca: Tocris Bioscience 6374/25
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
PF 04457845 is a potent and selective irreversible FAAH inhibitor (IC50 = 7.2 nM, which is selective for FAAH over a panel of other serine hydrolases. PF 04457845 exhibits efficacy in a rat inflammatory pain model, and is orally bioavailable and brain penetrant.Specifica
| N-3-Pyridazinyl-4-[[3-[[5-(trifluoromethyl)-2-pyridinyl]oxy]phenyl]methylene]-1-piperidinecarboxamide | |
| 1020315-31-4 | |
| >98% | |
| 25 mg |