missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ PF 06409577
Description
PF 06409577 is a potent and selective allosteric activator of AMPK (EC50 values are 7 and >40000 nM for AMPKα1β1γ1 and AMPKα1β2γ1, respectively). Exhibits minimal off-target effects on a panel of receptors, ion channels, PDEs and kinases. Improves kidney function and reduces proteinuria in a preclinical model of diabetic nephropathy. Orally bioavailable.
Specifications
Specifications
| Materiale o nome chimico | PF 06409577 |
| CAS | 1467057-23-3 |
| Sinonimo | 6-Chloro-5-[4-(1-hydroxycyclobutyl)phenyl]-1H-indole-3-carboxylic acid |
| Quantità | 50 mg |
| Solubilità | Soluble to 100mM in DMSO and to 20mM in ethanol |
| Linea di prodotti | Tocris |
| Formula molecolare | C19H16ClNO3 |
| Peso formulazione | 341.79 |
| Forma fisica | Solid |
| Colore | Gray |
| Show More |
Esclusivamente per scopi di ricerca.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?