missing translation for 'onlineSavingsMsg'
Learn More

PGBD3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Codice prodotto. p-7208181 Sfoglia Tutto Proteintech Prodotti
Cambia vista
Click to view available options
Quantità:
20 μL
150 μL
Dimensione della confezione:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
16897574 20 μL 20µL
16887574 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 16897574 Fornitore Proteintech N. del fornitore 176391AP20UL

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Rabbit Polyclonal Antibody

The piggyBac family of proteins, found in diverse animals, are transposases related to the transposase of the canonical piggyBac transposon from the moth, Trichoplusia ni. This family also includes genes in several genomes, including human, that appear to have been derived from the piggyBac transposons. Anti-PGBD3 belongs to the subfamily of piggyBac transposable element derived (PGBD) genes. The PGBD proteins appear to be novel, with no obvious relationship to other transposases, or other known protein families. Anti-PGBD3 overlaps with the ERCC6 gene on chromosome 10, and pseudogenes of this locus have been found on chromosomes 4, 5 and 12. The piggyBac family of proteins, found in diverse animals, are transposases related to the transposase of the canonical piggyBac transposon from the moth, Trichoplusia ni. This family also includes genes in several genomes, including human, that appear to have been derived from the piggyBac transposons. This gene belongs to the subfamily of piggyBac transposable element derived (PGBD) genes. The PGBD proteins appear to be novel, with no obvious relationship to other transposases, or other known protein families. This gene overlaps with the ERCC6 gene on chromosome 10, and pseudogenes of this locus have been found on chromosomes 4, 5 and 12.

Specifications

Antigene PGBD3
Applicazioni Western Blot, Immunohistochemistry (Paraffin)
Classificazione Polyclonal
Concentrazione 0.3 mg/mL
Coniugato Unconjugated
Formulazione PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PGBD3
N. accesso geni Q8N328
Alias gene PGBD3
Simboli geni Ercc6, PGBD3
Specie ospite Rabbit
Immunogeno PGBD3 Fusion Protein Ag11766
Metodo di purificazione Antigen Affinity Chromatography
Quantità 20 μL
Status giuridico RUO
Primario o secondario Primary
ID gene (immissione) 267004, 306274, 319955
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Forma Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.