missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ pH Sensor InLab™ Pure Pro-DES™
GREENER_CHOICE

Codice prodotto. 31454054 Sfoglia Tutto Mettler Toledo Prodotti
Cambia vista
Click to view available options
Tipi di connettore:
Direct
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Tipi di connettore
31454054 Direct
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 31454054 Fornitore Mettler Toledo™ N. del fornitore 31027066
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

A glass body, combination pH electrode with integrated temperature sensor, DES sensor head, bridge electrolyte and a fixed glass sleeve junction. Designed for low-ion samples like ultra-pure water, deionized water

  • Stable reading in low-ion samples: The high electrolyte flow permitted by the fixed sleeve junction reduces the potential at the junction, which in turn supports more stable readings
  • Always the right temperature: The integrated temperature sensor guarantees measurements are always performed with the correct temperature compensation
  • DES™ - Diagnosable Efficient Sustainable: Increased efficiency with sensor diagnostics: faster problem solving, less downtime, maximized sensor lifespan, and support of preventive maintenance.
  • Stay ahead: The sensor's diagnostic capability means peace of mind for the sensor condition. It supports preventive maintenance, maximizes the sensor's lifespan, and prevents premature discard
  • Sustainable investment: DES™ digital sensors are a smart choice. Retaining the measurement electronics and only replacing the sensor shaft at the end of its life, or the detachable cable that can be used for all sensors, helps reduce material waste and thus, the ecological footprint
  • Boost productivity and efficiency: With diagnosed errors, the user guidance with troubleshooting recommendations allows for faster problem-solving, thus reducing downtime. It also minimizes handling errors therefore cutting down on the need for specialized training
TRUSTED_SUSTAINABILITY

Specifica

Tipi di connettore Direct
Giunzione Immovable Glass Sleeve
Materiale Glass
Intervallo di pH 1 to 11
Tipo Combination Electrode/Integrated Temperature Probe
Riferimento interno ARGENTHAL™ with Ag+-trap
Membrana LoT
Sonda NTC 30 kΩ
Elettrolita di riferimento 1 mol/L KCl, 3 mol/L KCl
Compatibilità campioni Distilled Water,Sterile Water,Water For Injection
Dimensione campione ≥20 mL
Intervallo di temperatura (sistema metrico) 0°C to 80°C
Parametro pH
Tipi di sensore Combined Electrode
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.