Learn More
Proteintech Phantom Dye Blue 516 Viability Dye
Phantom Dye Blue 516 is a fixable viability dye which reacts with protein amine groups in both dead and live cells.
Marca:  Proteintech PD00007-100TESTS
Codice prodotto. 17358252
  • 60.00€ / 100 tests
 Articolo soggetto a restrizioni
Spedizione stimata: 24-11-2026
Aggiungi al carrello
Questo articolo non è restituibile. Consulta la politica di reso
Descrizione
Descrizione
  • Due to the compromised cell membrane in dead cells the dye can bind to many more proteins inside the cell, resulting in much higher fluorescence and easy differentiation of Live/Dead cells.
  • Phantom Dye Blue 516 is a water soluble violet fluorescent dye excited by the 488 nm laser making it an excellent addition to your multicolor panel.