Description
Magnetic head for the Pharma KingFisher™ Flex Deep-Well Head Magnetic Particle Processor.
This magnetic head can serve as a replacement head for the 96 deep-well format system. Includes one magnetic head.
Specifications
Specifications
| Linea di prodotti | KingFisher™ Flex |
| Quantità | 1 Head |
| Shipping Condition | Room Temperature |