Learn More
Esculape Laboratoire™ Pharmacy Cabinets
Product Code. 11718423
Change view
Click to view available options
Product Type:
Pharmacy Cabinet - Empty
Pharmacy Cabinet - Filled
Height (Metric):
14 cm
20 cm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Height (Metric) Unit Size
11718423 Pharmacy Cabinet - Empty 14 cm Each
11738423 Pharmacy Cabinet - Filled 20 cm Each
11708423 Pharmacy Cabinet - Filled 14 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11718423 Supplier Esculape Laboratoire™ Supplier No. 7110104
  • ONLINE EXCLUSIVE PRICE
    105.00€
    57.78€ / pezzi 1
    Risparmia 47.22€
    44% Sconto
Spedizione stimata: 01-12-2026
Specifications
Depth (Metric) 45 cm
For Use With (Application) To accommodate all the first aid equipment, offices, workshops and laboratories
Length (Metric) 30 cm
Product Type Pharmacy Cabinet - Empty
Quantity 1 Ea.
Height (Metric) 14 cm