Learn More
Phenanthroline, Tocris Bioscience™
Enhances hPSC differentiation into cranial placode cells
Marca: Tocris Bioscience 6385/100
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Phenanthroline is a enhances hPSC differentiation into cranial placode cells. Induces a 5-fold increase in cells expressing SIX1.Specifica
| 1,10-Phenanthroline | |
| 66-71-7 | |
| >99% | |
| 100 mg |