missing translation for 'onlineSavingsMsg'
Learn More

Phenetole, 98+%, Thermo Scientific Chemicals

Catalog Number p-7022632
Click to view available options
Quantità:
100 mL
500 mL
2500 mL
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
103-73-1
C8H10O
122.167
MFCD00009090
DLRJIFUOBPOJNS-UHFFFAOYSA-N
phenetole, benzene, ethoxy, ethyl phenyl ether, phenetol, phenyl ethyl ether, phenoxyethane, a phenoxyethane, ether, ethyl phenyl, unii-rb8lu2c57f
7674
CHEBI:67129
ethoxybenzene
CCOC1=CC=CC=C1
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
103-73-1
C8H10O
122.167
MFCD00009090
DLRJIFUOBPOJNS-UHFFFAOYSA-N
phenetole, benzene, ethoxy, ethyl phenyl ether, phenetol, phenyl ethyl ether, phenoxyethane, a phenoxyethane, ether, ethyl phenyl, unii-rb8lu2c57f
7674
CHEBI:67129
ethoxybenzene
CCOC1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico Phenetole
Punto di fusione -30°C
Densità 0.967
Punti di ebollizione 169°C to 170°C
Punto d'infiammabilità 57°C (134°F)
Odore Aromatic
Indice di rifrazione 1.507
Quantità 100 mL
Numero UN UN3271
Beilstein 636270
Indice di Merck 14,7229
Peso formulazione 122.17
Percent Purity ≥98%
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'