Learn More
R&D Systems™ Phenyl-glutarimide 4'-piperazine
Product Code. 30000416
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30000416 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30000416 Supplier R&D Systems™ Supplier No. 7729/100
 Restricted Item
Spedizione stimata: 27-11-2026
A Tocris product, part of R&D Systems. Piperazine-functionalized cereblon ligand for PROTAC development
Phenyl-glutarimide 4'-piperazine is a piperazine-functionalized cereblon ligand that can be used for PROTAC™ Degrader development. The compound can be used to develop PG-PROTACs that exhibit improved stability to hydrolysis and potency compared with PROTACs synthesized using IMiD (immunomodulatory imide drug) analogs.
Please contact us for SD files of our available Degrader Building Blocks.
PROTAC™ is a registered trademark of Arvinas Operations, Inc., and is used under license.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3-[4-(1-Piperazinyl)phenyl]-2,6-piperidinedione dihydrochloride
CAS 2902651-89-0
Quantity 100 mg
Product Line Tocris
Target Degrader Building Blocks
Molecular Formula C15H19N3O2.2HCl
Purity 95%