missing translation for 'onlineSavingsMsg'
Learn More

Phospho-MEK1/MEK2 (Ser217, Ser221) Recombinant Rabbit Monoclonal Antibody (MEK12S217S221-H2), PE, Invitrogen™

Rabbit Recombinant Monoclonal Antibody

Marca:  Thermo Scientific MA537344

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 17106938

  • 185.00€ / pezzi 10
Spedizione stimata: 28-05-2024
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

Phospho-MEK1/MEK2 (Ser217, Ser221) Recombinant Monoclonal antibody specifically detects Phospho-MEK1/MEK2 (Ser217, Ser221) in Human samples. It is validated for Flow Cytometry.
Specifica

Specifica

Phospho-MEK1/MEK2 (Ser217, Ser221)
Monoclonal
PE
MAP2K2
AA589381; CFC3; CFC4; dual specificity mitogen activated protein kinase kinase 1; dual specificity mitogen-activated protein kinase kinase 1; Dual specificity mitogen-activated protein kinase kinase 2; EC 2.7.12.2; ERK activator kinase 1; ERK activator kinase 2; hypothetical protein; hypothetical protein LOC406728; kinase MEK1; MAP kinase kinase 1; MAP kinase kinase 2; MAP kinase/Erk kinase; MAP kinase/Erk kinase 1; Map2k1; Map2k2; MAPK/ERK kinase 1; MAPK/ERK kinase 2; MAPKK 1; MAPKK 2; MAPKK1; MAPKK2; MEK 1; MEK 2; Mek1; MEK-1; Mek2; MEK-2; MEKK1; mitogen activated protein kinase kinase 1; mitogen activated protein kinase kinase 2; mitogen-activated protein kinase kinase 1; mitogen-activated protein kinase kinase 2; mitogen-activated protein kinase kinase 2, p45; mitogen-activated protein kinase kinase type 2; MK2; MKK1; MKK2; MP2K1; PRKMK1; PRKMK2; protein kinase, mitogen activated, kinase 1, p45; protein kinase, mitogen activated, kinase 2, p45; protein kinase, mitogen-activated, ki
Rabbit
Protein A/G
RUO
5604, 5605
4° C
Liquid
Flow Cytometry
MEK12S217S221-H2
PBS with 0.20% BSA and 0.09% sodium azide
P36507, Q02750
MAP2K1, MAP2K2
A synthetic phospho-peptide corresponding to residues surrounding Ser217/221 of human phospho MEK1/2.
10 Tests
Primary
Human
Antibody
IgG κ
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali