missing translation for 'onlineSavingsMsg'
Learn More

Phospho-p38 MAPK alpha (Thr180, Tyr182) Rabbit anti-Human, Mouse, Rat, APC, Clone: P38T180Y182-E3, Invitrogen™

Rabbit Monoclonal Antibody

Marca:  Thermo Scientific MA536914

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 17106508

  • 424.00€ / pezzi 100

Spedizione stimata: 31-10-2022
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

Phospho-p38 MAPK alpha (Thr180, Tyr182) Monoclonal antibody specifically detects Phospho-p38 MAPK alpha (Thr180, Tyr182) in Human, Mouse, Rat samples. It is validated for Flow Cytometry
Specifica

Specifica

Phospho-p38 MAPK alpha (Thr180, Tyr182)
Monoclonal
APC
MAPK14
186F5S; anon-sts23; BG:DS00797.3; CG7393; CG7393 gene product from transcript CG7393-RA; CG7393-PA; CRK1; CSAID-binding protein; Csaids binding protein; CSBP; CSBP1; CSBP2; CSPB1; cytokine suppressive anti-inflammatory drug binding protein; cytokine suppressive anti-inflammatory drug binding protein 1; cytokine suppressive anti-inflammatory drug-binding protein; cytokine-supressive anti-inflammatory drug binding protein; Dm p38b; Dmel\CG7393; Dmel_CG7393; Dmp38b; Dp38; D-p38; D-p38 MAPK; dp38b; D-p38b; EC 2.7.11.24; ERK1; ERT2; ESTS:186F5S; etID18993.8; EXIP; fk28c03; hm:zeh1243; Hog; I79_016605; MAP 14; MAP kinase 1; MAP kinase 14; MAP kinase 14A; MAP kinase 2; MAP kinase MXI2; MAP kinase p38; MAP kinase p38 alpha; MAP kinase p38a; MAP kinase p38alpha; MAP kinase p38b; MAPK; MAPK 14; MAPK 14A; MAPK1 antibody; MAPK14; mapk14a; MAX-interacting protein 2; mitogen activated protein kinase 14; mitogen-activated protein kinase 14; Mitogen-activated protein kinase 14A; mitogen-activated protein kinase p38 alpha; Mitogen-activated protein kinase p38a; Mitogen-activated protein kinase p38b; MK14; Mpk34C; Mxi2; OTTHUMP00000216563; OTTHUMP00000216566; p38; p38 alpha; p38 alpha/Mxi2; p38 beta; p38 MAP kinase; p38 MAP kinase alpha; p38 MAPK; p38 mitogen activated protein kinase; p38 mitogen-activated protein kinase; p38a; p38alpha; p38-alpha; p38alpha Exip; p38b; p38b MAP kinase; p38beta; p38b-PA; p38Hog; p38Kb; p38MAPK; Prkm14; PRKM15; reactive kinase; RK; RP1-179N16.5; SAPK2A; stress-activated p38b MAP kinase; stress-activated protein kinase 2A; tRNA synthetase cofactor p38; wu:fk28c03; zeh1243; zp38a
A synthetic phospho-peptide corresponding to residues surrounding Thr180/Tyr182 of human phospho p38 MAPK.
100 Tests
Primary
Rat, Mouse, Human
Antibody
IgG1 κ
Flow Cytometry
P38T180Y182-E3
PBS with 0.20% BSA and 0.09% sodium azide
P47811, P70618, Q16539
Rabbit
Protein A/G
RUO
1432, 26416, 81649
2-8°C
Liquid
Videos
SDS
Documenti
Certificati
Offerte speciali

Offerte speciali