Learn More
Phospho-p70(S6K) (Thr421/Ser424) Rabbit Polyclonal Antibody, Proteintech
Codice prodotto. 30233925 Sfoglia Tutto Proteintech Prodotti
Cambia vista
Click to view available options
Quantità:
20 μL
100 μL
Dimensione della confezione:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
30233925 20 μL 20µL
30237770 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Codice prodotto. 30233925 Fornitore Proteintech N. del fornitore 292481AP20UL
Rabbit Polyclonal Antibody
The Phospho-p70(S6K) (Thr421/Ser424) antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human Phospho-p70(S6K) (Thr421/Ser424). This antibody recognizes human antigen. The Phospho-p70(S6K) (Thr421/Ser424) antibody has been validated for the following applications: WB, IF/ICC, ELISA analysis.
Specifica
Antigene p70(S6K) (Thr421/Ser424)
Applicazioni Western Blot, Immunocytochemistry/Immunofluorescence, ELISA
Classificazione Polyclonal
Concentrazione 600 μg/mL
Coniugato Unconjugated
Simboli geni PS6K; S6K; S6K1; STK14A; p70(S6K)-alpha; p70-S6K; p70-alpha
Specie ospite Rabbit
Immunogeno Peptide
Metodo di purificazione Antigen affinity purification
Quantità 20 μL
Status giuridico RUO
Primario o secondario Primary
ID gene (immissione) 6198
Target Species Human
Content And Storage Store at -20°C. Stable for one year after shipment.
Antibody Molecular Weight 65-85 kDa
Product Type Polyclonal Antibody
Forma Liquid
Isotype IgG