missing translation for 'onlineSavingsMsg'
Learn More

Phospho-PTEN (Ser380, Thr382, Thr383) Rabbit anti-Human, Mouse, FITC, Clone: PTENS380T382383-E4, Invitrogen™

Rabbit Monoclonal Antibody

Marca:  Thermo Scientific MA536995

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 17116588

  • 424.00€ / pezzi 100

Spedizione stimata: 13-01-2023
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

Phospho-PTEN (Ser380, Thr382, Thr383) Monoclonal antibody specifically detects Phospho-PTEN (Ser380, Thr382, Thr383) in Human, Mouse samples. It is validated for Flow Cytometry
Specifica

Specifica

Phospho-PTEN (Ser380, Thr382, Thr383)
Monoclonal
FITC
PTEN
10q23del; 2310035O07Rik; A130070J02Rik; AI463227; B430203M17Rik; BZS; CWS1; DEC; GLM2; homolog of human mutated in multiple advanced cancers; MHAM; mitochondrial phosphatase and tensin protein alpha; mitochondrial PTENalpha; Mmac; MMAC1; MMAC1 phosphatase and tensin homolog deleted on chromosome 10; Mutated in multiple advanced cancers 1; phosphatase and tensin homolog; phosphatase and tensin homolog (mutated in multiple advanced cancers 1); phosphatase and tensin homolog deleted on chromosome ten; phosphatase and tensin-like protein; phosphatidylinositol 3; Phosphatidylinositol 3,4,5-trisphosphate 3-phosphatase and dual-specificity protein phosphatase PTEN; phosphatidylinositol-3; phosphatidylinositol-3,4,5-trisphosphate 3-phosphatase and dual-specificity protein phosphatase PTEN; protein tyrosine phosphatase and tensin-like protein; Pten; pten tumor suppressor; PTEN1; TEP1
A synthetic phospho-peptide corresponding to residues surrounding Ser380 and Thr382/383 of human phospho PTEN.
100 Tests
Primary
Mouse, Human
Antibody
IgG1 κ
Flow Cytometry
PTENS380T382383-E4
PBS with 0.20% BSA and 0.09% sodium azide
O08586, P60484
Rabbit
Protein A/G
RUO
19211, 5728
4° C, store in dark
Liquid
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali